Tisk…

8-(Trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid hydrate

Dodavatel: Apollo Scientific
Varování

PC300575-1G PC300575-250MG
APOSPC300575-1GEA 4300 CZK
APOSPC300575-1G APOSPC300575-250MG
8-(Trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid hydrate
8-(Trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid hydrate
Vzorec: C₉H₅F₃N₂O₂·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD22682817
CAS číslo: 1018828-72-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR