Perfluoro-15-crown-5-ether 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Icosafluoro-15-crown 5-Ether, Icosafluoro-1,4,7,10,13-pentaoxacyclopentadecane, Perfluoro-1,4,7,10,13-pentaoxacyclopentadecane, Perfluoro-15-crown 5-Ether
PC3005-1G
PC3005-5G
PC3005-1KG
PC3005-250G
PC3005-500G
PC3005-750G
APOSPC3005-1GEA
2280
CZK
APOSPC3005-1G
APOSPC3005-5G
APOSPC3005-1KG
APOSPC3005-250G
APOSPC3005-500G
APOSPC3005-750G
Perfluoro-15-crown-5-ether 95%
Perfluoro-15-crown-5-ether
Potential 19F NMR imaging agent, particularly effective in brain and spinal diagnostics, USP 4,838,274. Single nmr signal; liquid at room temperature.
Vzorec:
C₁₀F₂₀O₅ MW: 580,08 g/mol Bod varu: 146 °C Hustota: 1,3 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD01320747 CAS číslo: 97571-69-2 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...