2-Chloro-5-(trifluoromethoxy)phenylboronic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(2-Chloro-5-(trifluoromethoxy)phenyl)boronic acid, 2-Chloro-5-(trifluoromethoxy)benzeneboronic acid
PC2997-1G
PC2997-25G
PC2997-5G
APOSPC2997-1GEA
622
CZK
APOSPC2997-1G
APOSPC2997-25G
APOSPC2997-5G
2-Chloro-5-(trifluoromethoxy)phenylboronic acid
2-Chloro-5-(trifluoromethoxy)phenylboronic acid
Vzorec:
C₇H₅BClF₃O₃ MW: 240,37 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06656267 CAS číslo: 1022922-16-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...