5-Acetyl-2-fluorophenylboronic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(5-Acetyl-2-fluorophenyl)boronic acid
PC2994-5G
PC2994-10G
APOSPC2994-5GEA
3740
CZK
APOSPC2994-5G
APOSPC2994-10G
5-Acetyl-2-fluorophenylboronic acid
5-Acetyl-2-fluorophenylboronic acid
Vzorec:
C₈H₈BFO₃ MW: 181,9567 g/mol Bod tání: 112…117 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06656263 CAS číslo: 870777-29-0 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...