Tisk…

4-Acetyl-3-fluorophenylboronic acid

Dodavatel: Apollo Scientific
Varování

PC2993-1G PC2993-500MG
APOSPC2993-1GEA 1160 CZK
APOSPC2993-1G APOSPC2993-500MG
4-Acetyl-3-fluorophenylboronic acid
4-Acetyl-3-fluorophenylboronic acid
Vzorec: C₈H₈BFO₃
MW: 181,9600 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06796422
CAS číslo: 481725-35-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR