4-Fluoro-o-toluic acid 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC2983-100G
PC2983-25G
PC2983-5G
APOSPC2983-100GEA
3060
CZK
APOSPC2983-100G
APOSPC2983-25G
APOSPC2983-5G
4-Fluoro-o-toluic acid 97%
4-Fluoro-o-toluic acid
Vzorec:
C₈H₇FO₂ Bod tání: 168…172 °C Storage Temperature: Ambient |
MDL Number:
MFCD04115880 CAS číslo: 321-21-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...