Perfluoro-3,6,9-trioxadecanoic acid 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC2972-25G
PC2972-5G
PC2972-1G
APOSPC2972-25GEA
8900
CZK
APOSPC2972-25G
APOSPC2972-5G
APOSPC2972-1G
Perfluoro-3,6,9-trioxadecanoic acid 98%
Perfluoro-3,6,9-trioxadecanoic acid
Vzorec:
C₇HF₁₃O₅ Bod varu: 170…171 °C Storage Temperature: Ambient |
MDL Number:
MFCD00156025 CAS číslo: 151772-59-7 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...