Tisk…

2-Bromo-6-fluoroanisole

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-Bromo-3-fluoro-2-methoxybenzene

PC2958-25G
APOSPC2958-25GEA 3640 CZK
APOSPC2958-25G
2-Bromo-6-fluoroanisole
2-Bromo-6-fluoroanisole
Vzorec: C₇H₆BrFO
MW: 205,03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04974125
CAS číslo: 845829-94-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR