Tisk…

Methyl 2-Fluoro-2-methylpropionate 95%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-Fluoro-2-methylpropionic acid methyl ester

PC2925-1G PC2925-5G
APOSPC2925-1GEA 622 CZK
APOSPC2925-1G APOSPC2925-5G
Methyl 2-Fluoro-2-methylpropionate 95%
Methyl 2-Fluoro-2-methylpropionate
Vzorec: C₅H₉FO₂
MW: 120,12 g/mol
Bod varu: 108…109 °C
Hustota: 1 g/cm³ (25 °C)
Bod vzplanutí: 22 °C
Storage Temperature: Ambient
MDL Number: MFCD03265444
CAS číslo: 338-76-1
UN: 3272
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR