Tisk…

2,2-Difluorosuccinic acid 95%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: α,α-Difluorosuccinic acid, 2,2-Difluoro-1,4-butanedioic acid

PC2890-1G PC2890-5G
APOSPC2890-1GEA 1990 CZK
APOSPC2890-1G APOSPC2890-5G
2,2-Difluorosuccinic acid 95%
2,2-Difluorosuccinic acid
Vzorec: C₄H₄F₂O₄
MW: 154,07 g/mol
Bod tání: 144…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00040970
CAS číslo: 665-31-6
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR