Tisk…

2,5-Difluoro-DL-phenylalanine

Dodavatel: Apollo Scientific

Synonyms: 2-Amino-3-(2,5-difluorophenyl)propanoic acid, H-DL-Phe(2,5-DiF)-OH

PC2874I-1G PC2874I-5G PC2874I-25G
APOSPC2874I-1GEA 2280 CZK
APOSPC2874I-1G APOSPC2874I-5G APOSPC2874I-25G
2,5-Difluoro-DL-phenylalanine
2,5-Difluoro-DL-phenylalanine
Vzorec: C₉H₉F₂NO₂
MW: 201,170 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00061303
CAS číslo: 32133-38-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR