Tisk…

2,3-Difluoro-DL-phenylalanine 97%

Dodavatel: Apollo Scientific
Varování

PC2874G-1G PC2874G-5G PC2874G-10G
APOSPC2874G-1GEA 2070 CZK
APOSPC2874G-1G APOSPC2874G-5G APOSPC2874G-10G
2,3-Difluoro-DL-phenylalanine 97%
2,3-Difluoro-DL-phenylalanine
Vzorec: C₉H₉F₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD00061301
CAS číslo: 236754-62-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR