(2,6-Difluorophenyl)acetic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2,6-Difluorophenylacetic acid
PC2874D-25G
PC2874D-5G
PC2874D-100G
APOSPC2874D-25GEA
1910
CZK
APOSPC2874D-25G
APOSPC2874D-5G
APOSPC2874D-100G
(2,6-Difluorophenyl)acetic acid 97%
(2,6-Difluorophenyl)acetic acid
Vzorec:
(C₆H₃F₂)CH₂COOH MW: 172.13 Bod varu: 259 Bod tání: 100…102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010001 CAS číslo: 85068-28-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...