Tisk…

2,6-Difluoro-3-nitrobenzonitrile 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC2863-25G PC2863-5G
APOSPC2863-25GEA 1240 CZK
APOSPC2863-25G APOSPC2863-5G
2,6-Difluoro-3-nitrobenzonitrile 98%
2,6-Difluoro-3-nitrobenzonitrile
Vzorec: C₇H₂F₂N₂O₂
MW: 184.1
Bod tání: 50…54 °C
Storage Temperature: Ambient
MDL Number: MFCD00042254
CAS číslo: 143879-77-0
UN: 3439
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR