4-(Difluoromethoxy)benzonitrile 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC2847O-5G
PC2847O-25G
PC2847O-50G
APOSPC2847O-5GEA
1540
CZK
APOSPC2847O-5G
APOSPC2847O-25G
APOSPC2847O-50G
4-(Difluoromethoxy)benzonitrile 98%
4-(Difluoromethoxy)benzonitrile
Vzorec:
C₈H₅F₂NO MW: 169,1282 g/mol Bod tání: 40…42 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00085006 CAS číslo: 90446-25-6 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...