Tisk…
2,6-Dibromo-4-fluorphenyl isocyanate 97%
Dodavatel: Apollo Scientific
Varování
PC28258-1G PC28258-25G PC28258-5G
APOSPC28258-1GEA 1240 CZK
APOSPC28258-1G APOSPC28258-25G APOSPC28258-5G
2,6-Dibromo-4-fluorphenyl isocyanate 97%
2,6-Dibromo-4-fluorphenyl isocyanate
When melted, may contain small amount of sediment.
Vzorec: C₇H₂Br₂FNO
MW: 294,91 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00037041
CAS číslo: 76393-18-5
UN: 2811
ADR: 6.1,III