(4-Fluoro-1-benzothiophen-2-yl)methanol 95%
Dodavatel: Apollo Scientific
PC28222-1G
PC28222-5G
APOSPC28222-1GEA
9560
CZK
APOSPC28222-1G
APOSPC28222-5G
(4-Fluoro-1-benzothiophen-2-yl)methanol 95%
(4-Fluoro-1-benzothiophen-2-yl)methanol
Vzorec:
C₉H₇FOS MW: 182.22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12032275 CAS číslo: 957059-61-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...