Tisk…

5-(Diethoxymethyl)-2-fluorobenzonitrile 97%

Dodavatel: Apollo Scientific

PC28216-1G PC28216-5G
APOSPC28216-1GEA 7670 CZK
APOSPC28216-1G APOSPC28216-5G
5-(Diethoxymethyl)-2-fluorobenzonitrile 97%
5-(Diethoxymethyl)-2-fluorobenzonitrile
Vzorec: C₁₂H₁₄FNO₂
Storage Temperature: Ambient
MDL Number: MFCD18904305
CAS číslo: 1176161-37-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR