Tisk…

Methyl amino(3-fluorophenyl)acetate hydrochloride 95%

Dodavatel: Apollo Scientific

PC28213-1G PC28213-5G
APOSPC28213-1GEA 9560 CZK
APOSPC28213-1G APOSPC28213-5G
Methyl amino(3-fluorophenyl)acetate hydrochloride 95%
Methyl amino(3-fluorophenyl)acetate hydrochloride
Vzorec: C₉H₁₁ClFNO₂
MW: 219.64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09152736
CAS číslo: 42718-21-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR