2-(4-Fluorophenyl)-2-oxoethyl acetate 95%
Dodavatel: Apollo Scientific
PC28212-1G
PC28212-5G
APOSPC28212-1GEA
5130
CZK
APOSPC28212-1G
APOSPC28212-5G
2-(4-Fluorophenyl)-2-oxoethyl acetate 95%
2-(4-Fluorophenyl)-2-oxoethyl acetate
Vzorec:
C₁₀H₉FO₃ Storage Temperature: Ambient |
MDL Number:
MFCD09152727 CAS číslo: 366-78-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...