2-(2-Chloro-4-fluorophenoxy)acetonitrile 95%
Dodavatel: Apollo Scientific
PC28211-1G
PC28211-5G
APOSPC28211-1GEA
7670
CZK
APOSPC28211-1G
APOSPC28211-5G
2-(2-Chloro-4-fluorophenoxy)acetonitrile 95%
2-(2-Chloro-4-fluorophenoxy)acetonitrile
Vzorec:
C₈H₅ClFNO MW: 185.59 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09152771 CAS číslo: 24115-19-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...