Tisk…

2-Fluoro-3-(furan-2-yl)-3-oxopropanenitrile 95%

Dodavatel: Apollo Scientific

PC28209-1G PC28209-5G
APOSPC28209-1GEA 9560 CZK
APOSPC28209-1G APOSPC28209-5G
2-Fluoro-3-(furan-2-yl)-3-oxopropanenitrile 95%
2-Fluoro-3-(furan-2-yl)-3-oxopropanenitrile
Vzorec: C₇H₄FNO₂
Storage Temperature: Ambient
MDL Number: MFCD18426520
CAS číslo: 1263063-16-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR