Tisk…

1-(6-Fluoro-2-pyridinyl)tetrahydro-4(1H)-pyridinone 95%

Dodavatel: Apollo Scientific

Synonyms: 1-(6-Fluoropyridin-2-yl)piperidin-4-one

PC28169-1G PC28169-5G
APOSPC28169-1GEA 7670 CZK
APOSPC28169-1G APOSPC28169-5G
1-(6-Fluoro-2-pyridinyl)tetrahydro-4(1H)-pyridinone 95%
1-(6-Fluoro-2-pyridinyl)tetrahydro-4(1H)-pyridinone
Vzorec: C₁₀H₁₁FN₂O
MW: 194,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00665213
CAS číslo: 339096-62-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR