Tisk…

2,2,2-Trifluoro-1-(1-methyl-1H-indol-3-yl)-1-ethanol 95%

Dodavatel: Apollo Scientific

Synonyms: 2,2,2-Trifluoro-1-(1-methyl-1H-indol-3-yl)ethan-1-ol

PC28148-1G PC28148-5G
APOSPC28148-1GEA 9560 CZK
APOSPC28148-1G APOSPC28148-5G
2,2,2-Trifluoro-1-(1-methyl-1H-indol-3-yl)-1-ethanol 95%
2,2,2-Trifluoro-1-(1-methyl-1H-indol-3-yl)-1-ethanol
Vzorec: C₁₁H₁₀F₃NO
MW: 229,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02571238
CAS číslo: 116757-91-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR