Tisk…

3-Amino-3-[4-(trifluoromethyl)phenyl]acrylonitrile 95%

Dodavatel: Apollo Scientific

Synonyms: (2Z)-3-Amino-3-[4-(trifluoromethyl)phenyl]prop-2-enenitrile

PC28144-1G PC28144-5G
APOSPC28144-1GEA 5130 CZK
APOSPC28144-1G APOSPC28144-5G
3-Amino-3-[4-(trifluoromethyl)phenyl]acrylonitrile 95%
3-Amino-3-[4-(trifluoromethyl)phenyl]acrylonitrile
Vzorec: C₁₀H₇F₃N₂
MW: 212,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01871638
CAS číslo: 71682-89-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR