Tisk…

8-Fluoro-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole 95%

Dodavatel: Apollo Scientific

PC28107-1G
APOSPC28107-1GEA 6390 CZK
APOSPC28107-1G
8-Fluoro-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole 95%
8-Fluoro-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole
Vzorec: C₁₁H₁₁FN₂
MW: 190,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00277692
CAS číslo: 39876-39-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR