Tisk…

2,2-Difluoroethyl acetate 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2,2-Difluoroethylacetate

PC2808-25G PC2808-5G PC2808-100G PC2808-1G
APOSPC2808-25GEA 10350 CZK
APOSPC2808-25G APOSPC2808-5G APOSPC2808-100G APOSPC2808-1G
2,2-Difluoroethyl acetate 97%
2,2-Difluoroethyl acetate
Vzorec: C₄H₆F₂O₂
MW: 124.09 g/mol
Bod varu: 105 °C
Hustota: 1.2025 g/cm³ (25 °C)
Bod vzplanutí: 22 °C
Storage Temperature: Ambient
MDL Number: MFCD00069133
CAS číslo: 1550-44-3
UN: 3272
ADR: 3 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR