Tisk…

3-Fluoro-5-methoxybenzyl bromide 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC2797-1G PC2797-500MG PC2797-5G
APOSPC2797-1GEA 1950 CZK
APOSPC2797-1G APOSPC2797-500MG APOSPC2797-5G
3-Fluoro-5-methoxybenzyl bromide 97%
3-Fluoro-5-methoxybenzyl bromide
Vzorec: C₈H₈BrFO
Storage Temperature: Ambient
MDL Number: MFCD08741378
CAS číslo: 914637-29-9
UN: 3265
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR