2-Fluorophenyl methyl sulphone
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
1-Fluoro-2-(methylsulphonyl)benzene
PC2795-1G
PC2795-25G
PC2795-5G
APOSPC2795-1GEA
622
CZK
APOSPC2795-1G
APOSPC2795-25G
APOSPC2795-5G
2-Fluorophenyl methyl sulphone
2-Fluorophenyl methyl sulphone
Vzorec:
C₇H₇FO₂S MW: 174,2 g/mol Bod tání: 48…52 °C Bod vzplanutí: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD03094199 CAS číslo: 654-47-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...