Tisk…

trans-3,5-Difluorocinnamic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: trans-3-(3,5-Difluorophenyl)acrylic acid

PC2766-25G PC2766-100G PC2766-5G
APOSPC2766-25GEA 1740 CZK
APOSPC2766-25G APOSPC2766-100G APOSPC2766-5G
trans-3,5-Difluorocinnamic acid 98%
trans-3,5-Difluorocinnamic acid
Vzorec: C₉H₆F₂O₂
MW: 184.14
Bod tání: 204…205 °C
Storage Temperature: Ambient
MDL Number: MFCD00010321
CAS číslo: 147700-58-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR