Tisk…

trans-2,6-Difluorocinnamic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: (E)-3-(2,6-Difluorophenyl)acrylic acid

PC2763-10G PC2763-25G PC2763-1G PC2763-5G PC2763-100G
APOSPC2763-10GEA 913 CZK
APOSPC2763-10G APOSPC2763-25G APOSPC2763-1G APOSPC2763-5G APOSPC2763-100G
trans-2,6-Difluorocinnamic acid 98%
trans-2,6-Difluorocinnamic acid
Vzorec: C₉H₆F₂O₂
MW: 184,14 g/mol
Bod tání: 124…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00010319
CAS číslo: 102082-89-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR