3,4-Difluorobenzyl alcohol 96%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(3,4-Difluorophenyl)methanol
PC2733-25G
PC2733-1G
APOSPC2733-25GEA
665
CZK
APOSPC2733-25G
APOSPC2733-1G
3,4-Difluorobenzyl alcohol 96%
3,4-Difluorobenzyl alcohol
Vzorec:
C₇H₆F₂O MW: 144,12 g/mol Hustota: 1,262 g/cm³ (25 °C) Bod vzplanutí: 97 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010628 CAS číslo: 85118-05-4 EINECS: 285-657-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...