2,6-Difluorobenzyl alcohol 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(2,6-Difluorophenyl)methanol
PC2732-10G
PC2732-25G
APOSPC2732-10GEA
622
CZK
APOSPC2732-10G
APOSPC2732-25G
2,6-Difluorobenzyl alcohol 98%
2,6-Difluorobenzyl alcohol
Vzorec:
C₇H₆F₂O MW: 144.12 Bod varu: 88 °C/14mmHg Hustota: 1.3 g/cm³ (25 °C) Bod vzplanutí: 88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004603 CAS číslo: 19064-18-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...