2,4-Difluorobenzyl alcohol 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2,4-Difluorobenzyl Alcohol, (2,4-Difluorophenyl)methanol
PC2730-25G
APOSPC2730-25GEA
1620
CZK
APOSPC2730-25G
2,4-Difluorobenzyl alcohol 98%
2,4-Difluorobenzyl alcohol
Vzorec:
C₇H₆F₂O MW: 144,12 g/mol Bod varu: 51…52 °C/1mmHg Hustota: 1,29 g/cm³ (25 °C) Bod vzplanutí: 90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009983 CAS číslo: 56456-47-4 EINECS: 260-191-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...