Tisk…

4-Chloro-2-(trifluoromethyl)benzoic acid 97%

Dodavatel: Apollo Scientific

Synonyms: 4-Chloro-α,α,α-trifluoro-o-toluic acid

PC2709-25G PC2709-5G PC2709-100G
APOSPC2709-25GEA 2650 CZK
APOSPC2709-25G APOSPC2709-5G APOSPC2709-100G
4-Chloro-2-(trifluoromethyl)benzoic acid 97%
4-Chloro-2-(trifluoromethyl)benzoic acid
Vzorec: C₈H₄ClF₃O₂
Bod tání: 108…110 °C
Storage Temperature: Ambient
MDL Number: MFCD03094162
CAS číslo: 142994-09-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR