4-Chloro-2-(trifluoromethyl)benzonitrile 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Chloro-2-cyanobenzotrifluoride
PC2708-1G
PC2708-5G
PC2708-25G
APOSPC2708-1GEA
622
CZK
APOSPC2708-1G
APOSPC2708-5G
APOSPC2708-25G
4-Chloro-2-(trifluoromethyl)benzonitrile 98%
4-Chloro-2-(trifluoromethyl)benzonitrile
Vzorec:
C₈H₃ClF₃N MW: 205,57 g/mol Bod varu: 109 °C/10mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00060671 CAS číslo: 320-41-2 UN: 3276 ADR: 6.1,III |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...