Tisk…

Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate

Dodavatel: Apollo Scientific
Varování

Synonyms: TFFH, N-((Dimethylamino)fluoromethylene)-N-methylmethanaminium hexafluorophosphate(V)

PC2706-25G PC2706-100G
APOSPC2706-25GEA 1820 CZK
APOSPC2706-25G APOSPC2706-100G
Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate
Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate

Fluorinating Reagent

Vzorec: C₅H₁₂F₇N₂P
MW: 264,13 g/mol
Bod tání: 104…109 °C
Storage Temperature: Ambient
MDL Number: MFCD02684443
CAS číslo: 164298-23-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR