Tisk…

1-Chloro-3-fluoro-2-iodobenzene

Dodavatel: Apollo Scientific
Varování

PC2704-1G PC2704-5G PC2704-10G
APOSPC2704-1GEA 705 CZK
APOSPC2704-1G APOSPC2704-5G APOSPC2704-10G
1-Chloro-3-fluoro-2-iodobenzene
1-Chloro-3-fluoro-2-iodobenzene
Vzorec: C₆H₃ClFI
MW: 256,4400 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08458092
CAS číslo: 127654-70-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR