3,4,5-Trifluoroiodobenzene
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1,2,3-Trifluoro-5-iodobenzene
PC2701-1G
PC2701-5G
PC2701-10G
APOSPC2701-1GEA
622
CZK
APOSPC2701-1G
APOSPC2701-5G
APOSPC2701-10G
3,4,5-Trifluoroiodobenzene
3,4,5-Trifluoroiodobenzene
Vzorec:
C₆H₂F₃I MW: 257,9800 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD03265259 CAS číslo: 170112-66-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...