3,3'-Difluorobenzophenone 99%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Bis(3-fluorophenyl)methanone, Bis(3-fluorophenyl) ketone
PC2693-1G
PC2693-25G
PC2693-5G
APOSPC2693-1GEA
622
CZK
APOSPC2693-1G
APOSPC2693-25G
APOSPC2693-5G
3,3'-Difluorobenzophenone 99%
3,3'-Difluorobenzophenone
Vzorec:
C₁₃H₈F₂O MW: 218,2 g/mol Bod tání: 58…61 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000337 CAS číslo: 345-70-0 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...