Tisk…

3-Fluoro-(4-methoxycarbonyl)phenylboronic acid 96%

Dodavatel: Apollo Scientific
Varování

Synonyms: (3-Fluoro-4-(methoxycarbonyl)phenyl)boronic acid, 3-Fluoro-4-(methoxycarbonyl)phenylboronic acid, 3-Fluoro-4-methoxycarbonylphenylboronic acid, 3-Fluoro-4-(methoxycarbonyl)benzeneboronic acid

PC2667-25G
APOSPC2667-25GEA 3270 CZK
APOSPC2667-25G
3-Fluoro-(4-methoxycarbonyl)phenylboronic acid 96%
3-Fluoro-(4-methoxycarbonyl)phenylboronic acid
Vzorec: C₈H₈BFO₄
MW: 197,96 g/mol
Bod tání: 180…184 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04115665
CAS číslo: 505083-04-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR