Tisk…

1-(Trifluoromethanesulphonyl)-1H-benzotriazole

Dodavatel: Apollo Scientific
Nebezpečí

PC2658-1G PC2658-5G
APOSPC2658-1GEA 6580 CZK
APOSPC2658-1G APOSPC2658-5G
1-(Trifluoromethanesulphonyl)-1H-benzotriazole
1-(Trifluoromethanesulphonyl)-1H-benzotriazole
Vzorec: C₇H₄F₃N₃O₂S
MW: 251.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00963686
CAS číslo: 117632-84-5
UN: 1759
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR