Tisk…

3-Bromo-5-(trifluoromethoxy)benzaldehyde 98%

Dodavatel: Apollo Scientific
Varování

PC2615-1G PC2615-5G PC2615-25G
APOSPC2615-1GEA 622 CZK
APOSPC2615-1G APOSPC2615-5G APOSPC2615-25G
3-Bromo-5-(trifluoromethoxy)benzaldehyde 98%
3-Bromo-5-(trifluoromethoxy)benzaldehyde
Vzorec: C₈H₄BrF₃O₂
MW: 269,01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04116040
CAS číslo: 886498-07-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR