Tisk…

3-Bromo-5-(trifluoromethoxy)benzamide

Dodavatel: Apollo Scientific

PC2611-10G PC2611-5G
APOSPC2611-10GEA 12530 CZK
APOSPC2611-10G APOSPC2611-5G
3-Bromo-5-(trifluoromethoxy)benzamide
3-Bromo-5-(trifluoromethoxy)benzamide
Vzorec: C₈H₅BrF₃NO₂
MW: 284.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08458075
CAS číslo: 914636-32-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR