Tisk…

3-Chloro-2,6-difluorobenzyl alcohol 96%

Dodavatel: Apollo Scientific

Synonyms: (3-Chloro-2,6-difluorophenyl)methanol

PC2599-25G PC2599-5G PC2599-1G
APOSPC2599-25GEA 7940 CZK
APOSPC2599-25G APOSPC2599-5G APOSPC2599-1G
3-Chloro-2,6-difluorobenzyl alcohol 96%
3-Chloro-2,6-difluorobenzyl alcohol
Vzorec: C₇H₅ClF₂O
MW: 178.57
Storage Temperature: Ambient
MDL Number: MFCD01631325
CAS číslo: 252004-35-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR