Tisk…

Ishikawa's reagent 90%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1-(Diethylamino)-1,1,2,3,3,3-hexafluoropropane, Hexafluoropropene diethylamine, N,N-Diethyl-1,1,2,3,3,3-hexafluoropropylamine

PC2567-100G PC2567-25G
APOSPC2567-100GEA 9130 CZK
APOSPC2567-100G APOSPC2567-25G
Ishikawa's reagent 90%
Ishikawa's reagent

Active fluorinating agent see: Ishikawa, Bull. Chem.Soc. Jpn., 1979, 52, 3377

Vzorec: C₇H₁₁F₆N
Bod varu: 56…57 °C
Hustota: 1,23 g/cm³ (25 °C)
Bod vzplanutí: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00054683
CAS číslo: 309-88-6
UN: 2920
ADR: 8+3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR