Tisk…

Diethyl hexafluoroglutarate 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Diethyl perfluoroglutarate

PC2566-25G PC2566-5G PC2566-100G
APOSPC2566-25GEA 13890 CZK
APOSPC2566-25G APOSPC2566-5G APOSPC2566-100G
Diethyl hexafluoroglutarate 98%
Diethyl hexafluoroglutarate
Vzorec: C₉H₁₀F₆O₄
MW: 296.17
Bod varu: 224 °C
Hustota: 1.344 g/cm³ (25 °C)
Bod vzplanutí: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00015156
CAS číslo: 424-40-8
EINECS: 207-035-2
UN: 1760
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR