Tisk…

1-Fluoro-4-(1,1,2,2-tetrafluoroethoxy)benzene

Dodavatel: Apollo Scientific
Varování

Synonyms: alpha,alpha,beta,beta,4-Pentafluorophenetole

PC2561-1G PC2561-5G
APOSPC2561-1GEA 1660 CZK
APOSPC2561-1G APOSPC2561-5G
1-Fluoro-4-(1,1,2,2-tetrafluoroethoxy)benzene
1-Fluoro-4-(1,1,2,2-tetrafluoroethoxy)benzene
Vzorec: C₈H₅F₅O
Storage Temperature: Ambient
CAS číslo: 887268-36-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR