4-Fluoroisoquinoline 95+%
Dodavatel: Apollo Scientific
PC250003-10G
PC250003-1G
PC250003-5G
APOSPC250003-10GEA
12530
CZK
APOSPC250003-10G
APOSPC250003-1G
APOSPC250003-5G
4-Fluoroisoquinoline 95+%
4-Fluoroisoquinoline
Vzorec:
C₉H₆FN MW: 147,15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06656128 CAS číslo: 394-67-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...