5-Bromo-2-(trifluoromethoxy)toluene 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC2457-5G
PC2457-1G
PC2457-250MG
APOSPC2457-5GEA
16370
CZK
APOSPC2457-5G
APOSPC2457-1G
APOSPC2457-250MG
5-Bromo-2-(trifluoromethoxy)toluene 95%
5-Bromo-2-(trifluoromethoxy)toluene
Vzorec:
C₈H₆BrF₃O MW: 255,04 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04972695 CAS číslo: 887268-26-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...